Assay Details
Gene Symbol: RGD1308923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108698.1 | NP_001102168.1 | 3 - 4 | 682 | 70 | |
XM_006239585.2 | XP_006239647.1 | 3 - 4 | 1363 | 70 | ||
GenBank mRNA | FQ234838.1 | - | 3 - 4 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
