Assay Details
Gene Symbol: Rpa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021582.1 | NP_067593.1 | 2 - 3 | 205 | 65 | |
GenBank mRNA | BC079180.1 | - | 2 - 3 | 205 | 65 | |
X98490.1 | - | 1 - 2 | 111 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
