Assay Details
Gene Symbol: Noc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033897.1 | NP_001029069.1 | 10 - 11 | 1440 | 73 | |
XM_006239584.2 | XP_006239646.1 | 11 - 12 | 1354 | 73 | ||
GenBank mRNA | BC101911.1 | - | 10 - 11 | 1440 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
