Assay Details
Gene Symbol: Noc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033897.1 | NP_001029069.1 | 12 - 13 | 1662 | 89 | |
XM_006239584.2 | XP_006239646.1 | 13 - 14 | 1576 | 89 | ||
GenBank mRNA | BC101911.1 | - | 12 - 13 | 1662 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
