Assay Details
Gene Symbol: Zcchc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109267.1 | NP_001102737.1 | 7 - 8 | 704 | 97 | |
XM_006238968.2 | XP_006239030.1 | 7 - 8 | 724 | 97 | ||
XM_006238969.2 | XP_006239031.1 | 6 - 7 | 660 | 97 | ||
XM_006238970.2 | XP_006239032.1 | 5 - 6 | 539 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
