Assay Details
Gene Symbol: Zcchc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109267.1 | NP_001102737.1 | 8 - 9 | 1611 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P615732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62745 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P615732 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P615732 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P615732 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P615732 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 96954 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44061 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10880120 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880117 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880116 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880121 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880114 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880118 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880115 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880119 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10872473 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10880113 | - | - |
Rat Genome 230 2.0 Array | 1395399_at | - | - |
Rat Genome 230 2.0 Array | 1395399_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1395399_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1395399_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500027674.rn.1 | TC0500003705.rn.1 | RTA TC |
Rat Expression Set 230 | 1395399_at | - | - |
Rat Expression Set 230 | 1395399_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10880113 | - | - |
Rat Genome U34 Set | rc_AI176701_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21239784 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372517 | - | - |
