Assay Details
Gene Symbol: Rsrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014193.2 | NP_001014215.2 | 1 - 2 | 163 | 92 | |
GenBank mRNA | BC085888.1 | - | 1 - 2 | 111 | 92 | |
CB811984.1 | - | 1 - 2 | 174 | 92 | ||
FQ230600.1 | - | 1 - 2 | 148 | 92 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC085888.1, NM_001014193.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
