Assay Details
Gene Symbol: Rsrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014193.2 | NP_001014215.2 | 3 - 4 | 906 | 68 | |
GenBank mRNA | FQ230600.1 | - | 3 - 4 | 891 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
