Assay Details
Gene Symbol: Arhgef16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127591.1 | NP_001121063.1 | 12 - 13 | 1777 | 71 | |
XM_008764385.1 | XP_008762607.1 | 12 - 13 | 2396 | 71 | ||
XM_008764386.1 | XP_008762608.1 | 12 - 13 | 2563 | 71 | ||
XM_008764387.1 | XP_008762609.1 | 12 - 13 | 1950 | 71 | ||
GenBank mRNA | BC166429.1 | - | 12 - 13 | 1777 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
