Assay Details
Gene Symbol: Arhgef16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127591.1 | NP_001121063.1 | 5 - 6 | 960 | 104 | |
XM_008764385.1 | XP_008762607.1 | 5 - 6 | 1579 | 104 | ||
XM_008764386.1 | XP_008762608.1 | 5 - 6 | 1746 | 104 | ||
XM_008764387.1 | XP_008762609.1 | 5 - 6 | 1133 | 104 | ||
GenBank mRNA | BC166429.1 | - | 5 - 6 | 960 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
