Assay Details
Gene Symbol: S100pbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134628.1 | NP_001128100.1 | 2 - 3 | 237 | 79 | |
XM_006238956.2 | XP_006239018.1 | 2 - 3 | 204 | 79 | ||
XM_006238957.2 | XP_006239019.1 | 2 - 3 | 508 | 79 | ||
XM_006238958.2 | XP_006239020.1 | 2 - 3 | 220 | 79 | ||
XM_006238961.2 | XP_006239023.1 | 2 - 3 | 204 | 79 | ||
XM_006238962.2 | XP_006239024.1 | 2 - 3 | 203 | 79 | ||
XM_006238963.2 | XP_006239025.1 | 2 - 3 | 205 | 79 | ||
XM_008764162.1 | XP_008762384.1 | 2 - 3 | 204 | 79 | ||
XM_008764163.1 | XP_008762385.1 | 2 - 3 | 203 | 79 | ||
XM_008764164.1 | XP_008762386.1 | 2 - 3 | 203 | 79 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001134628.1,XM_006238956.2,XM_006238957.2,XM_006238958.2,XM_006238961.2,XM_006238962.2,XM_006238963.2,XM_008764162.1,XM_008764163.1,XM_008764164.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
