Assay Details
Gene Symbol: S100pbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134628.1 | NP_001128100.1 | 5 - 6 | 1202 | 149 | |
XM_006238956.2 | XP_006239018.1 | 5 - 6 | 1169 | 149 | ||
XM_006238957.2 | XP_006239019.1 | 5 - 6 | 1473 | 149 | ||
XM_006238958.2 | XP_006239020.1 | 5 - 6 | 1185 | 149 | ||
XM_006238959.2 | XP_006239021.1 | 4 - 5 | 1055 | 149 | ||
XM_006238960.1 | XP_006239022.1 | 4 - 5 | 1114 | 149 | ||
XM_006238961.2 | XP_006239023.1 | 6 - 7 | 1043 | 149 | ||
XM_006238963.2 | XP_006239025.1 | 6 - 7 | 939 | 149 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
