Assay Details
Gene Symbol: Gabre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023091.1 | NP_075579.1 | 3 - 4 | 1810 | 66 | |
GenBank mRNA | AF189262.1 | - | 3 - 4 | 1810 | 66 | |
AF255385.1 | - | 1372 | 66 | |||
AF255386.1 | - | 1 - 2 | 65 | 66 | ||
AF255387.1 | - | 1 - 2 | 65 | 66 | ||
AF255612.1 | - | 375 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
