Assay Details
Gene Symbol: Gabrq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031733.1 | NP_113921.1 | 2 - 3 | 222 | 145 | |
XM_006229516.2 | XP_006229578.1 | 3 - 4 | 386 | 145 | ||
GenBank mRNA | AF419333.1 | - | 2 - 3 | 222 | 145 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
