Assay Details
Gene Symbol: Gabra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017069.3 | NP_058765.3 | 5 - 6 | 786 | 110 | |
XM_006229511.2 | XP_006229573.1 | 5 - 6 | 898 | 110 | ||
GenBank mRNA | CB556618.1 | - | 4 - 5 | 614 | 110 | |
X51991.1 | - | 5 - 6 | 793 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
