Assay Details
Gene Symbol: Lsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013421.1 | NP_001013439.1 | 11 - 12 | 1554 | 62 | |
GenBank mRNA | BC079013.1 | - | 10 - 11 | 1400 | 62 | |
BC091338.1 | - | 11 - 12 | 1554 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
