Assay Details
Gene Symbol: Uqcrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004250.2 | NP_001004250.1 | 5 - 6 | 630 | 99 | |
GenBank mRNA | BC078923.1 | - | 5 - 6 | 632 | 99 | |
FQ213124.1 | - | 5 - 6 | 637 | 99 | ||
FQ215592.1 | - | 1 - 2 | 114 | 99 | ||
FQ216746.1 | - | 5 - 6 | 633 | 99 | ||
FQ224157.1 | - | 5 - 6 | 638 | 99 | ||
FQ229052.1 | - | 5 - 6 | 653 | 99 | ||
FQ231552.1 | - | 5 - 6 | 652 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
