Assay Details
Gene Symbol: Impdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199099.2 | NP_954530.1 | 10 - 11 | 1200 | 76 | |
XM_006243751.2 | XP_006243813.1 | 11 - 12 | 1313 | 76 | ||
GenBank mRNA | BC060585.1 | - | 10 - 11 | 1201 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
