Assay Details
Gene Symbol: Hyal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172040.2 | NP_742037.2 | 3 - 4 | 1159 | 87 | |
XM_006243855.2 | XP_006243917.1 | 3 - 4 | 1180 | 87 | ||
GenBank mRNA | AF535141.1 | - | 2 - 3 | 1067 | 87 | |
BC062410.1 | - | 3 - 4 | 1159 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
