Assay Details
Gene Symbol: Hhatl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106868.1 | NP_001100338.1 | 1 - 2 | 144 | 123 | |
XM_006244091.2 | XP_006244153.1 | 1 - 2 | 225 | 123 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106868.1, XM_006244091.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
