Assay Details
Gene Symbol: Vcam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012889.1 | NP_037021.1 | 712 | 80 | ||
GenBank mRNA | BC089812.1 | - | 730 | 80 | ||
M84488.1 | - | 712 | 80 | |||
X63722.1 | - | 661 | 80 |
Gene Symbol: LOC100912479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749384.3 | XP_003749432.1 | 3 - 4 | 925 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
