Assay Details
Gene Symbol: Dnajb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013076.1 | NP_001013094.1 | 3 - 4 | 366 | 79 | |
XM_006233488.1 | XP_006233550.1 | 2 - 3 | 348 | 79 | ||
GenBank mRNA | BC083638.1 | - | 3 - 4 | 366 | 7134832 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC083638.1, NM_001013076.1, XM_006233488.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
