Assay Details
Gene Symbol: Gipc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037210.1 | NP_001032287.1 | 2 - 3 | 444 | 83 | |
XM_008761552.1 | XP_008759774.1 | 2 - 3 | 1670 | 83 | ||
GenBank mRNA | BC100257.1 | - | 2 - 3 | 444 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
