Assay Details
Gene Symbol: Gipc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037210.1 | NP_001032287.1 | 5 - 6 | 813 | 80 | |
GenBank mRNA | BC100257.1 | - | 5 - 6 | 813 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P114500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781147 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177739 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P114500 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P114500 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P114500 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P306932 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P114500 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29471 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62948 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 75298 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35700 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38392 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10940668 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10940668 | - | - |
Rat Genome 230 2.0 Array | 1383287_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383287_at | - | - |
Rat Genome 230 2.0 Array | 1389354_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389354_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200033858.rn.1 | TC0200004901.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200033857.rn.1 | TC0200004901.rn.1 | RTA TC |
Rat Expression Set 230 | 1389354_PM_at | - | - |
Rat Expression Set 230 | 1389354_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10940668 | - | - |
Rat Genome U34 Set | rc_AI103787_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21421122 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351859 | - | - |
