Assay Details
Gene Symbol: Myoz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106469.1 | NP_001099939.1 | 1 - 2 | 130 | 88 | |
XM_006233264.2 | XP_006233326.1 | 1 - 2 | 175 | 88 | ||
GenBank mRNA | FQ215232.1 | - | 1 - 2 | 70 | 88 | |
FQ215285.1 | - | 1 - 2 | 100 | 88 | ||
FQ215331.1 | - | 1 - 2 | 129 | 88 | ||
FQ215936.1 | - | 1 - 2 | 70 | 88 | ||
FQ216608.1 | - | 1 - 2 | 148 | 88 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001106469.1, XM_006233264.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
