Assay Details
Gene Symbol: Myoz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233264.2 | XP_006233326.1 | 2 - 3 | 270 | 66 | |
GenBank mRNA | FQ215232.1 | - | 2 - 3 | 165 | 66 | |
FQ215285.1 | - | 2 - 3 | 195 | 66 | ||
FQ215331.1 | - | 2 - 3 | 224 | 66 | ||
FQ215936.1 | - | 2 - 3 | 165 | 66 | ||
FQ216537.1 | - | 2 - 3 | 226 | 66 | ||
FQ216608.1 | - | 2 - 3 | 243 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
