Assay Details
Gene Symbol: Dram2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025018.1 | NP_001020189.1 | 7 - 8 | 861 | 93 | |
XM_006233120.2 | XP_006233182.1 | 7 - 8 | 805 | 93 | ||
XM_006233121.2 | XP_006233183.1 | 6 - 7 | 745 | 93 | ||
XM_006233122.2 | XP_006233184.1 | 6 - 7 | 846 | 93 | ||
GenBank mRNA | BC091251.1 | - | 7 - 8 | 861 | 93 | |
FQ218349.1 | - | 7 - 8 | 873 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
