Assay Details
Gene Symbol: Dram2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025018.1 | NP_001020189.1 | 2 - 3 | 203 | 143 | |
XM_006233120.2 | XP_006233182.1 | 2 - 3 | 147 | 143 | ||
XM_006233123.2 | XP_006233185.1 | 2 - 3 | 149 | 143 | ||
GenBank mRNA | BC091251.1 | - | 2 - 3 | 203 | 7372838 | 143 |
FQ218349.1 | - | 2 - 3 | 212 | 143 | ||
FQ228782.1 | - | 2 - 3 | 300 | 143 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC091251.1, NM_001025018.1, XM_006233120.2, XM_006233123.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
