Assay Details
Gene Symbol: Sars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007606.1 | NP_001007607.1 | 1 - 2 | 200 | 97 | |
XM_006233136.2 | XP_006233198.1 | 1 - 2 | 235 | 97 | ||
GenBank mRNA | BC061765.1 | - | 1 - 2 | 200 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
