Assay Details
Gene Symbol: Zzz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134549.2 | NP_001128021.2 | 4 - 5 | 1951 | 75 | |
XM_006233489.2 | XP_006233551.1 | 6 - 7 | 2235 | 75 | ||
XM_006233490.2 | XP_006233552.1 | 7 - 8 | 2825 | 75 | ||
XM_006233491.2 | XP_006233553.1 | 6 - 7 | 2232 | 75 | ||
XM_008761551.1 | XP_008759773.1 | 7 - 8 | 2264 | 75 | ||
GenBank mRNA | FM103952.1 | - | 2 - 3 | 140 | 75 | |
FN806367.1 | - | 1 - 2 | 114 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
