Assay Details
Gene Symbol: Adh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126120.1 | NP_001119592.1 | 3 - 4 | 270 | 76 | |
GenBank mRNA | BC083724.1 | - | 3 - 4 | 270 | 76 | |
FQ212950.1 | - | 3 - 4 | 326 | 76 | ||
FQ213560.1 | - | 3 - 4 | 315 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
