Assay Details
Gene Symbol: Hadh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057186.2 | NP_476534.1 | 5 - 6 | 665 | 104 | |
GenBank mRNA | AF095449.1 | - | 5 - 6 | 665 | 104 | |
FQ212781.1 | - | 5 - 6 | 645 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
