Assay Details
Gene Symbol: Cryz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012183.2 | NP_001012183.1 | 7 - 8 | 807 | 101 | |
XM_006233520.2 | XP_006233582.1 | 8 - 9 | 1043 | 101 | ||
GenBank mRNA | BC078927.1 | - | 7 - 8 | 809 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
