Assay Details
Gene Symbol: Slc6a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033079.1 | NP_001028251.1 | 11 - 12 | 2291 | 110 | |
GenBank mRNA | L06434.2 | - | 11 - 12 | 2291 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
