Assay Details
Gene Symbol: Slc6a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033079.1 | NP_001028251.1 | 6 - 7 | 1343 | 93 | |
GenBank mRNA | L06434.2 | - | 6 - 7 | 1343 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
