Assay Details
Gene Symbol: Papss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106471.1 | NP_001099941.1 | 4 - 5 | 600 | 77 | |
GenBank mRNA | BC166793.1 | - | 4 - 5 | 673 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
