Assay Details
Gene Symbol: Papss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106471.1 | NP_001099941.1 | 5 - 6 | 718 | 75 | |
GenBank mRNA | BC166793.1 | - | 5 - 6 | 791 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
