Assay Details
Gene Symbol: Hs2st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100518.1 | NP_001093988.1 | 6 - 7 | 1123 | 67 | |
GenBank mRNA | AY724508.1 | - | 6 - 7 | 1123 | 67 | |
BC091291.1 | - | 4 - 5 | 479 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P091053 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P775635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8543 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533600 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937473 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122739 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99600 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136863 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P091053 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P775635 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P091053 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P775635 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P775635 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P091053 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P541317 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102916 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42943 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69075 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32262 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72157 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66386 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10827133 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827130 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827136 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827137 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827134 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827135 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827131 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827132 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10827129 | - | - |
Rat Genome 230 2.0 Array | 1382101_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383724_PM_at | - | - |
Rat Genome 230 2.0 Array | 1382101_at | - | - |
Rat Genome 230 2.0 Array | 1380235_at | - | - |
Rat Genome 230 2.0 Array | 1383724_at | - | - |
Rat Genome 230 2.0 Array | 1380235_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382101_at | - | - |
Affymetrix HT Rat Focus Array | 1380235_at | - | - |
Affymetrix HT Rat Focus Array | 1380235_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382101_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200033478.rn.1 | TC0200004843.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200033479.rn.1 | TC0200004843.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200033480.rn.1 | TC0200004843.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200033481.rn.1 | TC0200004843.rn.1 | RTA TC |
Rat Expression Set 230 | 1383724_PM_at | - | - |
Rat Expression Set 230 | 1380235_PM_at | - | - |
Rat Expression Set 230 | 1380235_at | - | - |
Rat Expression Set 230 | 1382101_PM_at | - | - |
Rat Expression Set 230 | 1382101_at | - | - |
Rat Expression Set 230 | 1383724_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827129 | - | - |
Rat Genome U34 Set | rc_AA924751_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363349 | - | - |
