Assay Details
Gene Symbol: Cd53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012523.2 | NP_036655.1 | 1 - 2 | 132 | 136 | |
GenBank mRNA | BC078900.1 | - | 1 - 2 | 83 | 136 | |
M57276.1 | - | 1 - 2 | 146 | 136 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC078900.1, M57276.1, NM_012523.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
