Assay Details
Gene Symbol: Cd53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012523.2 | NP_036655.1 | 6 - 7 | 648 | 62 | |
GenBank mRNA | BC078900.1 | - | 6 - 7 | 599 | 62 | |
M57276.1 | - | 6 - 7 | 662 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
