Assay Details
Gene Symbol: Gnat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108950.2 | NP_001102420.2 | 6 - 7 | 675 | 93 | |
XM_006233171.1 | XP_006233233.1 | 6 - 7 | 708 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
