Assay Details
Gene Symbol: Tspan5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004090.2 | NP_001004090.2 | 1 - 2 | 87 | 94 | |
GenBank mRNA | AF540877.1 | - | 1 - 2 | 113 | 94 | |
BC100135.1 | - | 1 - 2 | 87 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
