Assay Details
Gene Symbol: Adh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134329.1 | NP_599156.1 | 3 - 4 | 316 | 80 | |
XM_008761493.1 | XP_008759715.1 | 4 - 5 | 703 | 80 | ||
GenBank mRNA | X98746.1 | - | 3 - 4 | 316 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
