Assay Details
Gene Symbol: Rpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100794.1 | NP_001094264.1 | 3 - 4 | 389 | 130 | |
GenBank mRNA | BC086515.1 | - | 3 - 4 | 358 | 130 | |
BC166888.1 | - | 3 - 4 | 386 | 130 | ||
BC166920.1 | - | 3 - 4 | 386 | 130 | ||
DY564389.1 | - | 3 - 4 | 368 | 130 | ||
FQ220191.1 | - | 3 - 4 | 368 | 130 | ||
FQ226064.1 | - | 3 - 4 | 375 | 130 | ||
FQ227364.1 | - | 3 - 4 | 375 | 130 | ||
FQ234387.1 | - | 3 - 4 | 375 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
