Assay Details
Gene Symbol: Rabggtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138708.2 | NP_619715.1 | 1 - 2 | 121 | 66 | |
GenBank mRNA | BC060536.1 | - | 145 | 66 | ||
BC078683.1 | - | 1 - 2 | 110 | 66 | ||
FQ214773.1 | - | 125 | 66 | |||
FQ216744.1 | - | 125 | 66 | |||
FQ229109.1 | - | 125 | 66 | |||
L10416.1 | - | 1 - 2 | 121 | 66 | ||
S62097.1 | - | 1 - 2 | 121 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
