Assay Details
Gene Symbol: Dnase2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021664.1 | NP_067696.1 | 5 - 6 | 894 | 95 | |
XM_006233464.2 | XP_006233526.1 | 6 - 7 | 971 | 95 | ||
XM_006233465.2 | XP_006233527.1 | 6 - 7 | 998 | 95 | ||
XM_006233466.2 | XP_006233528.1 | 6 - 7 | 1001 | 95 | ||
XM_006233468.2 | XP_006233530.1 | 6 - 7 | 1382 | 95 | ||
XM_008761521.1 | XP_008759743.1 | 6 - 7 | 1104 | 95 | ||
GenBank mRNA | AF178974.1 | - | 5 - 6 | 894 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
