Assay Details
Gene Symbol: Zranb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031616.1 | NP_113804.1 | 6 - 7 | 531 | 116 | |
GenBank mRNA | AF013965.1 | - | 6 - 7 | 771 | 116 | |
AF013967.1 | - | 6 - 7 | 531 | 116 | ||
BC087012.1 | - | 6 - 7 | 517 | 7189430 | 116 | |
FQ211774.1 | - | 6 - 7 | 557 | 116 | ||
FQ211917.1 | - | 6 - 7 | 557 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
