Assay Details
Gene Symbol: Frrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761474.1 | XP_008759696.1 | 15 - 16 | 1889 | 91 | |
GenBank mRNA | FQ210266.1 | - | 15 - 16 | 1826 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
