Assay Details
Gene Symbol: Slc25a24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127544.1 | NP_001121016.1 | 2 - 3 | 376 | 143 | |
GenBank mRNA | BC162022.1 | - | 2 - 3 | 355 | 143 | |
FQ221591.1 | - | 2 - 3 | 456 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
