Assay Details
Gene Symbol: Slc25a24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127544.1 | NP_001121016.1 | 5 - 6 | 729 | 76 | |
XM_008761404.1 | XP_008759626.1 | 4 - 5 | 757 | 76 | ||
GenBank mRNA | BC162022.1 | - | 5 - 6 | 708 | 76 | |
FQ211892.1 | - | 4 - 5 | 689 | 76 | ||
FQ221591.1 | - | 5 - 6 | 809 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
